α-Ethyl-4-methoxy-α,3,5-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166838-1G 166838-500MG 166838-5G 166838-25G
77465-564EA 341.96 USD
77465-564 77478-320 77488-504 77499-114
α-Ethyl-4-methoxy-α,3,5-trimethylbenzenemethanol ≥95%
α-Ethyl-4-methoxy-α,3,5-trimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD20528395
CAS Number: 1443354-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR