α-Ethyl-α,3,5-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

166790-1G 166790-500MG 166790-5G
77465-560EA 648.9 USD
77465-560 77478-316 77488-500
α-Ethyl-α,3,5-trimethylbenzenemethanol ≥95%
α-Ethyl-α,3,5-trimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12806371
CAS Number: 854179-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR