1-Methyl-3-(propylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

166721-1G 166721-500MG 166721-5G 166721-25G
77465-552EA 177.16 USD
77465-552 77478-308 77488-492 77499-110
1-Methyl-3-(propylthio)benzene ≥95%
1-Methyl-3-(propylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26142714
CAS Number: 24767-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR