4-(Butylthio)-1,2-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

166687-1G 166687-500MG 166687-5G 166687-25G
77465-548EA 415.09 USD
77465-548 77478-304 77488-486 77499-104
4-(Butylthio)-1,2-dichlorobenzene ≥97%
4-(Butylthio)-1,2-dichlorobenzene

Precaución: For research use only

Formula: C₁₀H₁₂Cl₂S
MW: 235.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34852015
CAS Number: 103753-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR