2-(tert-Butyl)phthalazin-1(2H)-one ≥95%

Supplier: AOBChem USA

Warning

166454-1G 166454-500MG 166454-5G 166454-25G
77465-540EA 195.7 USD
77465-540 77478-298 77488-478 77499-098
2-(tert-Butyl)phthalazin-1(2H)-one ≥95%
2-(tert-Butyl)phthalazin-1(2H)-one

Precaución: For research use only

Formula: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD32068184
CAS Number: 1167442-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR