3-(4-Ethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166372-1G 166372-500MG 166372-5G
77465-536EA 533.54 USD
77465-536 77478-294 77488-474
3-(4-Ethylphenyl)-3-oxetanol ≥95%
3-(4-Ethylphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851916
CAS Number: 1504489-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR