3-(4-Methoxy-3,5-dimethylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166361-1G 166361-500MG 166361-5G
77465-532EA 740.57 USD
77465-532 77478-290 77488-470
3-(4-Methoxy-3,5-dimethylphenyl)-3-oxetanol ≥95%
3-(4-Methoxy-3,5-dimethylphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851906
CAS Number: 1888958-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR