3-[4-(Methylthio)phenyl]-3-oxetanol ≥97%

Supplier: AOBChem USA

Warning

166349-1G 166349-500MG 166349-5G 166349-25G
77465-528EA 369.77 USD
77465-528 77478-286 77488-466 77499-094
3-[4-(Methylthio)phenyl]-3-oxetanol ≥97%
3-[4-(Methylthio)phenyl]-3-oxetanol

Precaución: For research use only

Formula: C₁₀H₁₂O₂S
MW: 196.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30627913
CAS Number: 1894507-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR