3-(3,5-Dimethoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166344-1G 166344-500MG 166344-5G
77465-524EA 541.78 USD
77465-524 77478-282 77488-462
3-(3,5-Dimethoxyphenyl)-3-oxetanol ≥95%
3-(3,5-Dimethoxyphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34851890
CAS Number: 1903713-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR