3-(2-Ethoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166332-1G 166332-500MG 166332-5G
77465-520EA 640.66 USD
77465-520 77478-278 77488-458
3-(2-Ethoxyphenyl)-3-oxetanol ≥95%
3-(2-Ethoxyphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34851878
CAS Number: 2104004-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR