3-[3-(Methylthio)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166296-1G 166296-500MG 166296-5G
77465-516EA 696.28 USD
77465-516 77478-274 77488-454
3-[3-(Methylthio)phenyl]-3-oxetanol ≥95%
3-[3-(Methylthio)phenyl]-3-oxetanol

Precaución: For research use only

Formula: C₁₀H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34851843
CAS Number: 2160762-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR