3-(2,4-Dimethylphenyl)-3-oxetanol ≥97%

Supplier: AOBChem USA

Warning

166281-1G 166281-500MG 166281-5G 166281-25G
77465-512EA 189.52 USD
77465-512 77478-270 77488-450 77499-090
3-(2,4-Dimethylphenyl)-3-oxetanol ≥97%
3-(2,4-Dimethylphenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34851829
CAS Number: 1401961-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR