3-[2-(1-Methylethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166266-1G 166266-500MG 166266-5G
77465-508EA 558.26 USD
77465-508 77478-266 77488-446
3-[2-(1-Methylethyl)phenyl]-3-oxetanol ≥95%
3-[2-(1-Methylethyl)phenyl]-3-oxetanol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34851816
CAS Number: 2110273-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR