3-(2-Naphthalenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166255-1G 166255-500MG 166255-5G
77465-504EA 434.66 USD
77465-504 77478-262 77488-442
3-(2-Naphthalenyl)-3-oxetanol ≥95%
3-(2-Naphthalenyl)-3-oxetanol

Precaución: For research use only

Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851807
CAS Number: 1896605-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR