3-Hydroxy-2,4-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16622-1G 16622-500MG 16622-5G
77465-500EA 609.76 USD
77465-500 77478-258 77488-438
3-Hydroxy-2,4-dimethoxyphenylboronic acid ≥97%
3-Hydroxy-2,4-dimethoxyphenylboronic acid

Precaución: For research use only

Formula: C₈H₁₁BO₅
MW: 197.981 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380491
CAS Number: 865540-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR