3-(2,6-Difluorophenyl)oxetan-3-ol ≥97%

Supplier: AOBChem USA

Warning

166214-1G 166214-500MG 166214-5G
77465-496EA 597.4 USD
77465-496 77478-254 77488-434
3-(2,6-Difluorophenyl)oxetan-3-ol ≥97%
3-(2,6-Difluorophenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851794
CAS Number: 2159796-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR