3-(2-Fluoro-5-methylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166208-1G 166208-500MG 166208-5G
77465-492EA 543.84 USD
77465-492 77478-250 77488-430
3-(2-Fluoro-5-methylphenyl)oxetan-3-ol ≥95%
3-(2-Fluoro-5-methylphenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.1915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851792
CAS Number: 2107690-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR