3-(4-Methoxyphenyl)oxetane ≥95%

Supplier: AOBChem USA

Warning

166201-1G 166201-500MG 166201-5G 166201-25G
77465-488EA 415.09 USD
77465-488 77478-246 77488-426 77499-084
3-(4-Methoxyphenyl)oxetane ≥95%
3-(4-Methoxyphenyl)oxetane

Precaución: For research use only

Formula: C₁₀H₁₂O₂
MW: 164.2010 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26127570
CAS Number: 1401878-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR