3-(4-Fluoro-2-methylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166193-1G 166193-500MG 166193-5G
77465-484EA 434.66 USD
77465-484 77478-242 77488-422
3-(4-Fluoro-2-methylphenyl)oxetan-3-ol ≥95%
3-(4-Fluoro-2-methylphenyl)oxetan-3-ol

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.1915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851789
CAS Number: 2107240-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR