6-Bromo-2,3-dimethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

16618-1G 16618-500MG 16618-5G 16618-25G
77465-480EA 183.34 USD
77465-480 77478-238 77488-418 77499-078
6-Bromo-2,3-dimethoxyphenol ≥97%
6-Bromo-2,3-dimethoxyphenol

Precaución: For research use only

Formula: C₈H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD13186752
CAS Number: 114605-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR