(3,6-Dichloro-2-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165936-1G 165936-500MG 165936-5G 165936-25G
77465-476EA 415.09 USD
77465-476 77478-234 77488-414 77499-074
(3,6-Dichloro-2-fluorophenyl)(ethyl)sulfane ≥95%
(3,6-Dichloro-2-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇Cl₂FS
Storage Temperature: Ambient
MDL Number: MFCD34851611
CAS Number: 2921858-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR