(6-Bromo-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165929-1G 165929-500MG 165929-5G 165929-25G
77465-472EA 415.09 USD
77465-472 77478-230 77488-410 77499-070
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFS₂
Storage Temperature: Ambient
MDL Number: MFCD34851606
CAS Number: 2921906-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR