(6-Bromo-2-fluoro-3-propoxyphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165920-1G 165920-500MG 165920-5G 165920-25G
77465-468EA 415.09 USD
77465-468 77478-226 77488-406 77499-066
(6-Bromo-2-fluoro-3-propoxyphenyl)(ethyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₄BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34851597
CAS Number: 2921798-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR