(3,6-Dibromo-2-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165914-1G 165914-500MG 165914-5G 165914-25G
77465-464EA 415.09 USD
77465-464 77478-222 77488-402 77499-062
(3,6-Dibromo-2-fluorophenyl)(ethyl)sulfane ≥95%
(3,6-Dibromo-2-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇Br₂FS
Storage Temperature: Ambient
MDL Number: MFCD34851591
CAS Number: 2921755-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR