(3-Bromo-2-chloro-6-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165911-1G 165911-500MG 165911-5G 165911-25G
77465-460EA 415.09 USD
77465-460 77478-218 77488-398 77499-058
(3-Bromo-2-chloro-6-fluorophenyl)(ethyl)sulfane ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇BrClFS
Storage Temperature: Ambient
MDL Number: MFCD34851588
CAS Number: 2921858-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR