(6-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165908-1G 165908-500MG 165908-5G 165908-25G
77465-456EA 415.09 USD
77465-456 77478-214 77488-394 77499-054
(6-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFS
Storage Temperature: Ambient
MDL Number: MFCD34851585
CAS Number: 2921735-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR