(2-Chloro-6-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165877-1G 165877-500MG 165877-5G 165877-25G
77465-452EA 415.09 USD
77465-452 77478-212 77488-390 77499-050
(2-Chloro-6-fluoro-3-methylphenyl)(ethyl)sulfane ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀ClFS
Storage Temperature: Ambient
MDL Number: MFCD34851558
CAS Number: 2921897-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR