(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165870-1G 165870-500MG 165870-5G 165870-25G
77465-448EA 415.09 USD
77465-448 77478-208 77488-386 77499-046
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(ethyl)sulfane ≥95%
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₂BrFS
Storage Temperature: Ambient
MDL Number: MFCD34851552
CAS Number: 2921867-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR