(E)-1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165836-1G 165836-500MG 165836-5G 165836-25G
77465-440EA 415.09 USD
77465-440 77478-200 77488-378 77499-038
(E)-1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)but-2-en-1-ol ≥95%
(E)-1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂BrFOS
MW: 291.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851521
CAS Number: 2921912-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR