(E)-1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165819-1G 165819-500MG 165819-5G 165819-25G
77465-428EA 415.09 USD
77465-428 77478-188 77488-366 77499-026
(E)-1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)but-2-en-1-ol ≥95%
(E)-1-(3-(benzyloxy)-2-bromo-6-fluorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₇H₁₆BrFO₂
MW: 351.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851504
CAS Number: 2921913-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR