(E)-1-(2-Chloro-6-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

165783-1G 165783-500MG 165783-5G 165783-25G
77465-420EA 415.09 USD
77465-420 77478-180 77488-358 77499-018
(E)-1-(2-Chloro-6-fluoro-3-methylphenyl)but-2-en-1-ol ≥95%
(E)-1-(2-Chloro-6-fluoro-3-methylphenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD34851469
CAS Number: 2921912-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR