3-Methoxyphenyl acetate ≥97%

Supplier: AOBChem USA

Warning

165201-1G 165201-500MG 165201-5G 165201-25G
77465-412EA 184.37 USD
77465-412 77478-172 77488-348 77499-010
3-Methoxyphenyl acetate ≥97%
3-Methoxyphenyl acetate

Precaución: For research use only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00026193
CAS Number: 5451-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR