4-Chloro-5-(methylthio)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

165181-1G 165181-500MG 165181-5G 165181-25G
77465-408EA 415.09 USD
77465-408 77478-168 77488-344 77499-006
4-Chloro-5-(methylthio)benzo[d][1,3]dioxole ≥97%
4-Chloro-5-(methylthio)benzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₈H₇ClO₂S
MW: 202.6579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34851054
CAS Number: 2921734-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR