2-(2-Bromo-6-chloro-3-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

16485-1G 16485-500MG 16485-5G 16485-25G
77465-400EA 188.49 USD
77465-400 77478-160 77488-336 77498-998
2-(2-Bromo-6-chloro-3-methylphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-6-chloro-3-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976875
CAS Number: 2221812-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR