3-(3-Bromo-2,6-difluorophenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164668-1G 164668-500MG 164668-5G
77465-392EA 447.02 USD
77465-392 77478-152 77488-328
3-(3-Bromo-2,6-difluorophenyl)pentan-3-ol ≥97%
3-(3-Bromo-2,6-difluorophenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34850800
CAS Number: 1695390-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR