3-(5-Bromo-2-fluorophenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164665-1G 164665-500MG 164665-5G
77465-388EA 447.02 USD
77465-388 77478-148 77488-324
3-(5-Bromo-2-fluorophenyl)pentan-3-ol ≥97%
3-(5-Bromo-2-fluorophenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD23759090
CAS Number: 1514816-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR