3-(3-(dimethylamino)phenyl)pentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

164663-1G 164663-500MG 164663-5G 164663-25G
77465-384EA 341.96 USD
77465-384 77478-144 77488-320 77498-988
3-(3-(dimethylamino)phenyl)pentan-3-ol ≥95%
3-(3-(dimethylamino)phenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₃H₂₁NO
MW: 207.3119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528644
CAS Number: 1443324-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR