3-(5-Fluoro-2-methoxyphenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164660-1G 164660-500MG 164660-5G 164660-25G
77465-380EA 341.96 USD
77465-380 77478-140 77488-316 77498-984
3-(5-Fluoro-2-methoxyphenyl)pentan-3-ol ≥97%
3-(5-Fluoro-2-methoxyphenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD20528628
CAS Number: 1443346-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR