3-(3-Fluoro-5-methoxyphenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164656-1G 164656-500MG 164656-5G 164656-25G
77465-376EA 397.58 USD
77465-376 77478-136 77488-312 77498-980
3-(3-Fluoro-5-methoxyphenyl)pentan-3-ol ≥97%
3-(3-Fluoro-5-methoxyphenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
MW: 212.2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850798
CAS Number: 2028500-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR