2-(5-Bromo-2-fluorophenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164639-1G 164639-500MG 164639-5G
77465-372EA 447.02 USD
77465-372 77478-132 77488-306
2-(5-Bromo-2-fluorophenyl)butan-2-ol ≥97%
2-(5-Bromo-2-fluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759089
CAS Number: 1533713-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR