2-(3-Fluoro-5-methoxyphenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164635-1G 164635-500MG 164635-5G 164635-25G
77465-368EA 397.58 USD
77465-368 77478-128 77488-302 77498-974
2-(3-Fluoro-5-methoxyphenyl)butan-2-ol ≥97%
2-(3-Fluoro-5-methoxyphenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO₂
MW: 198.2340 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850796
CAS Number: 1934578-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR