2-(3-Bromophenyl)butan-2-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
164625-1G
164625-500MG
164625-5G
164625-25G
77465-364EA
297.67
USD
77465-364
77478-124
77488-298
77498-970
2-(3-Bromophenyl)butan-2-ol ≥97%
2-(3-Bromophenyl)butan-2-ol
Precaución: For research use only
Formula:
C₁₀H₁₃BrO MW: 229.1136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12068092 CAS Number: 58977-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...