2-(4-Fluorophenyl)butan-2-ol ≥97%

Supplier: AOBChem USA

Warning

164622-1G 164622-500MG 164622-5G 164622-25G
77465-360EA 415.09 USD
77465-360 77478-120 77488-294 77498-966
2-(4-Fluorophenyl)butan-2-ol ≥97%
2-(4-Fluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₃FO
MW: 168.2080 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12067971
CAS Number: 7119-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR