1-(2-Fluorophenyl)pentan-1-ol ≥96%

Supplier: AOBChem USA

Warning

164583-1G 164583-500MG 164583-5G
77465-356EA 471.74 USD
77465-356 77478-116 77488-288
1-(2-Fluorophenyl)pentan-1-ol ≥96%
1-(2-Fluorophenyl)pentan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD17288667
CAS Number: 478076-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR