2-phenylphthalazin-1(2H)-one ≥95%

Supplier: AOBChem USA

Warning

164579-1G 164579-500MG 164579-5G 164579-25G
77465-352EA 168.92 USD
77465-352 77478-112 77488-284 77498-960
2-phenylphthalazin-1(2H)-one ≥95%
2-phenylphthalazin-1(2H)-one

Precaución: For research use only

Formula: C₁₄H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00278209
CAS Number: 6266-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR