1-(4-(dimethylamino)phenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

164577-1G 164577-500MG 164577-5G 164577-25G
77465-348EA 341.96 USD
77465-348 77478-108 77488-280 77498-956
1-(4-(dimethylamino)phenyl)butan-1-ol ≥95%
1-(4-(dimethylamino)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD11521093
CAS Number: 861532-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR