(4-Bromophenyl)(butyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164565-1G 164565-5G 164565-25G
77465-340EA 64.89 USD
77465-340 77488-272 77498-950
(4-Bromophenyl)(butyl)sulfane ≥97%
(4-Bromophenyl)(butyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃BrS
Storage Temperature: Ambient
MDL Number: MFCD12546517
CAS Number: 121392-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR