Butyl(4-chlorophenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164561-1G 164561-500MG 164561-5G 164561-25G
77465-336EA 262.65 USD
77465-336 77478-098 77488-268 77498-946
Butyl(4-chlorophenyl)sulfane ≥97%
Butyl(4-chlorophenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃ClS
MW: 200.7282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04274939
CAS Number: 16155-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR