(3-Bromo-2-fluorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

164559-1G 164559-500MG 164559-5G 164559-25G
77465-332EA 415.09 USD
77465-332 77478-094 77488-264 77498-942
(3-Bromo-2-fluorophenyl)(propyl)sulfane ≥95%
(3-Bromo-2-fluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFS
MW: 249.1431 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850781
CAS Number: 2921683-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR