(2,5-Difluorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

164556-1G 164556-500MG 164556-5G
77465-328EA 696.28 USD
77465-328 77478-090 77488-260
(2,5-Difluorophenyl)(propyl)sulfane ≥95%
(2,5-Difluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂S
Storage Temperature: Ambient
MDL Number: MFCD34850778
CAS Number: 1862516-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR